Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Piperidinepropionic acid, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
26371-07-3
C8H15NO2
157.21
LPDGWMLCUHULJF-UHFFFAOYSA-N
1-piperidinepropionic acid,1-piperidinepropanoic acid,3-piperidin-1-yl propanoic acid,unii-6l69a7vpho,piperidine-1-propionic acid,3-1-piperidinyl propanoic acid,3-piperidinopropanoic acid,6l69a7vpho,piperidinepropionic acid,3-1-piperidinyl propionic acid
117782
3-piperidin-1-ylpropanoic acid
C1CCN(CC1)CCC(=O)O
26371-07-3
C8H15NO2
157.21
LPDGWMLCUHULJF-UHFFFAOYSA-N
1-piperidinepropionic acid,1-piperidinepropanoic acid,3-piperidin-1-yl propanoic acid,unii-6l69a7vpho,piperidine-1-propionic acid,3-1-piperidinyl propanoic acid,3-piperidinopropanoic acid,6l69a7vpho,piperidinepropionic acid,3-1-piperidinyl propionic acid
117782
3-piperidin-1-ylpropanoic acid
C1CCN(CC1)CCC(=O)O
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 157.21
Physical Form Powder
Percent Purity 96%
Color Brown to White
Melting Point 86.0°C to 88.5°C
Quantity 25g
Chemical Name or Material 1-Piperidinepropionic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

EINECSNumber : 247-633-