Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanesulfonic acid, sodium salt, 98%, Acros Organics

Click to view available options
Quantity:
1g
14533-63-2
C3H7NaO3S
146.136
MFCD00062546
NPAWNPCNZAPTKA-UHFFFAOYSA-M
sodium 1-propanesulfonate,sodium propanesulphonate,1-propanesulfonic acid sodium salt,sodium propane-1-sulfonate,sodium propanesulfonate,acmc-209cuo,ipc-alks-3,c3h7o3s.na,potassium propane-1-sulfonate,propanesulfonic acid sodium salt
4319363
sodium;propane-1-sulfonate
CCCS(=O)(=O)[O-].[Na+]
14533-63-2
C3H7NaO3S
146.136
MFCD00062546
NPAWNPCNZAPTKA-UHFFFAOYSA-M
sodium 1-propanesulfonate,sodium propanesulphonate,1-propanesulfonic acid sodium salt,sodium propane-1-sulfonate,sodium propanesulfonate,acmc-209cuo,ipc-alks-3,c3h7o3s.na,potassium propane-1-sulfonate,propanesulfonic acid sodium salt
4319363
sodium;propane-1-sulfonate
CCCS(=O)(=O)[O-].[Na+]
Quantity 1g
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 146.14
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 1-Propanesulfonic acid, sodium salt, 98%

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 238-564-7

TSCA : TSCA