Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanethiol 98%, ACROS Organics™

Click to view available options
Quantity:
50mL
500mL
Packaging:
Glass bottle
107-03-9
C3H8S
76.157
MFCD00004900
SUVIGLJNEAMWEG-UHFFFAOYSA-N
1-propanethiol,propanethiol,n-propylmercaptan,propyl mercaptan,n-propyl mercaptan,n-propylthiol,1-propylmercaptan,propylthiol,1-mercaptopropane,1-propyl mercaptan
7848
CHEBI:8473
propane-1-thiol
CCCS
107-03-9
C3H8S
76.157
MFCD00004900
SUVIGLJNEAMWEG-UHFFFAOYSA-N
1-propanethiol,propanethiol,n-propylmercaptan,propyl mercaptan,n-propyl mercaptan,n-propylthiol,1-propylmercaptan,propylthiol,1-mercaptopropane,1-propyl mercaptan
7848
CHEBI:8473
propane-1-thiol
CCCS
Color Colorless
Infrared Spectrum Authentic
Density 0.84g/mL
Quantity 500mL
Packaging Glass bottle
Refractive Index 1.4370 to 1.4390 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3CH2CH2SH
Beilstein 01, 359
Formula Weight 76.15
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1-Propanethiol, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-455-5

RTECSNumber : TZ7300000

TSCA : TSCA