Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, ACS, >99.5%, Alfa Aesar™

Click to view available options
Quantity:
500mL
1L
4L
4 x 4L
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
Assay Percent Range >99.5%
Beilstein 1098242
Solubility Information Miscible with water,alcohols,ketones,aldehydes,ethers,glycols,aromatic and aliphatic hydrocarbons.
Formula Weight 60.1
Physical Form Liquid
Grade ACS
Density 0.804 g/cm3 at 20°C
Ignition Point 824°C (1515°F)
Odor Alcohol-like
Refractive Index 1.385
Vapor Pressure 10mm Hg at 14°C
Flash Point 15°C (59°F)
Boiling Point 97°C
Color Colorless
Melting Point °C to 127°C
Quantity 500mL
Chemical Name or Material 1-Propanol, In ACS analysis
Show More Show Less

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c

H225-H318-H336

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 200-746-9

RTECSNumber : UH8225000

TSCA : Yes

Recommended Storage : Ambient temperatures