Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Alfa Aesar™

Click to view available options
Quantity:
250mL
1L
4 x 1L
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
Assay Percent Range 99.9%
Appearance Clear liquid
Beilstein 1098242
Solubility Information Completely soluble in water. Miscible with propylene glycol.
Formula Weight 60.1
Physical Form Liquid
Density 0.804
Ignition Point 824°C (1515°F)
Odor Alcohol-like
Refractive Index 1.385
Vapor Pressure 13.3 hPa (10mm Hg) at 20°C
Flash Point 15°C (59°F)
Boiling Point 97°C
Color Colorless
Melting Point °C to 127°C
Quantity 250mL
Chemical Name or Material 1-Propanol
Show More Show Less

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P370+P378q-P501c

H225-H318-H336

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 200-746-9

RTECSNumber : UH8225000

TSCA : Yes

Recommended Storage : Ambient temperatures