Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
Poly Bottle

Chemical Identifiers

71-23-8
C3H8O
60.096
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
CHEBI:28831
propan-1-ol
CCCO

High quality chemicals for laboratory and specialized industrial use

Formula Weight 60.1
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 0.80kg
Flash Point 23.00°C
Quantity 4L
Chemical Name or Material 1-Propanol
Grade BAKER ANALYZED™ Reagent