Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
Poly Bottle

Chemical Identifiers

71-23-8
C3H8O
60.096
BDERNNFJNOPAEC-UHFFFAOYSA-N
1031
CHEBI:28831

High quality chemicals for laboratory and specialized industrial use

CAS 71-23-8
Molecular Formula C3H8O
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
SMILES CCCO
IUPAC Name propan-1-ol
Molecular Weight (g/mol) 60.096
PubChem CID 1031
ChEBI CHEBI:28831
Formula Weight 60.1
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Flash Point 23.00°C
Quantity 500mL
Chemical Name or Material 1-Propanol
Grade BAKER ANALYZED™ Reagent
Show More Show Less