Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
Poly Bottle
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
71-23-8
C3H8O
60.10
MFCD00002941
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO

High quality chemicals for laboratory and specialized industrial use

Formula Weight 60.1
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.80kg
Flash Point 23.00°C
Quantity 500mL
Chemical Name or Material 1-Propanol