1-Propanol (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Steel Pail
Quantity 20L

 View more versions of this product

Catalog No. A414-20

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 71-23-8
    Molecular Formula C3H8O
    Linear Formula CH3CH2CH2OH
    Formula Weight 60.09
    MDL Number MFCD00002941
    Synonym n-Propyl Alcohol
    Chemical Name or Material 1-Propanol
    Grade Certified Grade

    Specifications

    Boiling Point 97°C
    Melting Point −127°C
    Formula Weight 60.09
    Quantity 20L
    DOT Information DOT Class 3, : Flammable Liquid
    Physical Form Liquid
    Color Colorless
    pH 7
    Density 0.800g/cm³
    Viscosity 2.2 mPa/s at 20°C
    Alpha Vector Propanol
    Vapor Pressure 25mbar at 20°C
    Health Hazard 3 Emergency Overview
    Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause skin and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

    NFPA

    Health:1
    Flammability:3
    Instability:0
    Identification Pass Test
    Synonym n-Propyl Alcohol
    Additional Information Vapor Pressure: 25mbar at 20°C
    Packaging Steel Pail
    Residue after Evaporation 0.01% max.
    Specific Gravity 0.801 to 0.803 (at 25°C)
    Cleanup Symbol Solvent
    MDL Number MFCD00002941
    ChemAlert Storage Symbol Red
    Boiling Range 2°C max.
    Acidity 0.02% max.
    Molecular Formula C3H8O
    Linear Formula CH3CH2CH2OH
    CAS 71-23-8
    Health Hazard 2 DANGER!
    Chemical Name or Material 1-Propanol
    Grade Certified Grade
       

    DANGER!

    Emergency Overview
    Flammable liquid and vapor. Vapors may cause drowsiness and dizziness. May cause skin and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

    NFPA

    Health:1
    Flammability:3
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit