1-Propanol, Spectrophotometric Grade, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 22933M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA22933M1

 N/A
    N/A

This unit of measure is invalid. 


    Chemical Identifiers

    CAS 71-23-8
    Physical Form Liquid
    Assay Percent Range 99%
    Molecular Formula CH3CH2CH2OH
    Formula Weight 60.1
    MDL Number MFCD00002941
    Chemical Name or Material 1-Propanol
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 60.1
    Quantity 4 x 1L
    Color Clear
    DOT Information DOT Class: 3; Packing Group: II
    EINECS Number 200-746-9
    Physical Form Liquid
    Odor Alcohol-like
    Vapor Pressure 13.3 hPa (10mm Hg) at 20°C
    Appearance Clear liquid
    Molecular Formula CH3CH2CH2OH
    MDL Number MFCD00002941
    Health Hazard 1 H225-H318-H336: Highly flammable liquid and vapour. Causes serious eye damage. May cause drowsiness or dizziness.
    Assay Percent Range 99%
    CAS 71-23-8
    Chemical Name or Material 1-Propanol
    Grade Spectrophotometric grade

    H225-H318-H336: Highly flammable liquid and vapour. Causes serious eye damage. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit