Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, for spectroscopy ACS, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
71-23-8
C3H8O
60.1
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
71-23-8
C3H8O
60.1
BDERNNFJNOPAEC-UHFFFAOYSA-N
1-propanol,propanol,propyl alcohol,n-propanol,n-propyl alcohol,ethylcarbinol,1-hydroxypropane,optal,osmosol extra,propylic alcohol
1031
CHEBI:28831
propan-1-ol
CCCO
For UV/VIS spectroscopy
Viscosity 2.2 mPa.s (20°C)
Titratable Acid 0.0004 meq/g
Boiling Point 97.0°C
Residue after Evaporation 0.001% max.
Specific Gravity 0.8
Water 0.2% max.
Packaging Glass bottle
Refractive Index 1.3840 to 1.3860
Quantity 1L
Melting Point -127.0°C
Color Undesignated
Flash Point 15°C
Density 0.8000g/mL
Infrared Spectrum Authentic
Linear Formula CH3CH2CH2OH
Merck Index 15,7955
Solubility Information Clear colorless
Formula Weight 60.1
Physical Form Liquid
Percent Purity ≥99.5%
Grade Spectroscopy ACS
Chemical Name or Material 1-Propanol, For Spectroscopy
Show More Show Less

GHS H Statement
May cause drowsiness or dizziness.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 200-746-9