Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
14092-04-7,109-99-9
C3H5BrMg
145.28
WQMNNHNWITXOAH-UHFFFAOYSA-M
1-propenylmagnesium bromide,propenylmagnesium bromide,1-propenyl magnesium bromide,magnesium, bromo-1z-1-propenyl,e-prop-1-en-1-ylmagnesium bromide,propenyl magnesium bromide,z-propenylmagnesium bromide,prop-1-enylmagnesium bromide,e-propenylmagnesium bromide,prop-1-en-1-ylmagnesium bromide
5112801
magnesium;prop-1-ene;bromide
CC=[CH-].[Mg+2].[Br-]
14092-04-7,109-99-9
C3H5BrMg
145.28
WQMNNHNWITXOAH-UHFFFAOYSA-M
1-propenylmagnesium bromide,propenylmagnesium bromide,1-propenyl magnesium bromide,magnesium, bromo-1z-1-propenyl,e-prop-1-en-1-ylmagnesium bromide,propenyl magnesium bromide,z-propenylmagnesium bromide,prop-1-enylmagnesium bromide,e-propenylmagnesium bromide,prop-1-en-1-ylmagnesium bromide
5112801
magnesium;prop-1-ene;bromide
CC=[CH-].[Mg+2].[Br-]
Packaging AcroSeal™ Glass Bottle
Flash Point −20°C
Boiling Point 65.0°C
Density 0.9500g/mL
Color Brown to Yellow
Specific Gravity 0.95
Quantity 100mL
CAS Min % 6.88,91.59
CAS Max % 8.41,93.12
Solubility Information Solubility in water: reacts.
Formula Weight 145.28
Physical Form Solution
Chemical Name or Material 1-Propenylmagnesium bromide
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently w

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger