Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propyl-4-piperidone, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
23133-37-1
C8H15NO
141.214
MFCD00010433
YGDZKYYCJUNORF-UHFFFAOYSA-N
1-propyl-4-piperidone,n-propyl-4-piperidone,1-propyl-piperidin-4-one,4-piperidinone, 1-propyl,1-propylpiperidin-4-on,1-propyl4-piperidone,pubchem2038,n-propyl-4-peperidone,1-n-propyl-4-piperidon,1-propyl-4-piperidinone
90006
1-propylpiperidin-4-one
CCCN1CCC(=O)CC1
23133-37-1
C8H15NO
141.214
MFCD00010433
YGDZKYYCJUNORF-UHFFFAOYSA-N
1-propyl-4-piperidone,n-propyl-4-piperidone,1-propyl-piperidin-4-one,4-piperidinone, 1-propyl,1-propylpiperidin-4-on,1-propyl4-piperidone,pubchem2038,n-propyl-4-peperidone,1-n-propyl-4-piperidon,1-propyl-4-piperidinone
90006
1-propylpiperidin-4-one
CCCN1CCC(=O)CC1
Density 0.9360g/mL
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 141.21
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1-Propyl-4-piperidone
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Recommended Storage : Gains color during storage. Product may gradually polymerize during storage