Alfa Aesar™ 1-Propylboronic acid pinacol ester, 98%

Manufacturer: Alfa Aesar™ H3044114

Quantity 25g

 View more versions of this product

Catalog No. AAH3044114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67562-19-0
    Molecular Formula C9H19BO2
    Formula Weight 170.06
    MDL Number MFCD12546187
    UN Number UN1993
    Synonym 4,4,5,5-Tetramethyl-2-n-propyl-1,3,2-dioxaborolane
    Chemical Name or Material 1-Propylboronic acid pinacol ester

    Specifications

    Formula Weight 170.06
    Molecular Formula C9H19BO2
    MDL Number MFCD12546187
    CAS 67562-19-0
    UN Number UN1993
    Synonym 4,4,5,5-Tetramethyl-2-n-propyl-1,3,2-dioxaborolane
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Boiling Point 166° to 168°C
    Flash Point 47°C (117°F)
    Quantity 25g
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 1-Propylboronic acid pinacol ester
    Assay 98%

    Description

    Applications
    It is employed as a intermediates for pharmaceutical.

    Solubility
    Not miscible or difficult to mix with water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit