Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Pyridin-3-ylmethyl)-1H-pyrazole-4-boronic acid pinacol ester, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
864754-21-2
C15H20BN3O2
285.16
LVLQAPQDEZOFQU-UHFFFAOYSA-N
3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl methyl pyridine,1-pyridin-3-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester,3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrazol-1-ylmethyl-pyridine,1-3-pyridylmethyl-1h-pyrazole-4-boronic acid pinacol ester,3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl methyl pyridine,3-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl methyl pyridine,pubchem18546,1-pyridin-3-ylmethyl-1h-pyrazole-4-boronic acid
17999185
3-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methyl]pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3=CN=CC=C3
864754-21-2
C15H20BN3O2
285.16
LVLQAPQDEZOFQU-UHFFFAOYSA-N
3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol-1-yl methyl pyridine,1-pyridin-3-ylmethyl-1h-pyrazole-4-boronic acid, pinacol ester,3-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrazol-1-ylmethyl-pyridine,1-3-pyridylmethyl-1h-pyrazole-4-boronic acid pinacol ester,3-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl methyl pyridine,3-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazol-1-yl methyl pyridine,pubchem18546,1-pyridin-3-ylmethyl-1h-pyrazole-4-boronic acid
17999185
3-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazol-1-yl]methyl]pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)CC3=CN=CC=C3
Quantity 5g
Melting Point 102.0°C to 106.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 285.16
Percent Purity 97%
Chemical Name or Material 1-(Pyridin-3-ylmethyl)-1H-pyrazole-4-boronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning