Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pyrrolidinebutanenitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
35543-25-0
C8H14N2
138.21
MFCD00003183
OSDDDHPYSNZBPF-UHFFFAOYSA-N
1-pyrrolidinebutanenitrile,4-pyrrolidin-1-yl butanenitrile,1-pyrrolidinebutyronitrile,1-3-cyanopropyl pyrrolidine,4-1-pyrrolidine butyronitrile,1-pyrrolidinobutyronitrile,4-pyrrolidin-1-yl-butyronitrile,4-pyrrolidinylbutanenitrile,4-pyrrolidin-1-ylbutyronitrile,4-pyrrolidinobutyronitrile
118838
4-pyrrolidin-1-ylbutanenitrile
C1CCN(C1)CCCC#N
35543-25-0
C8H14N2
138.21
MFCD00003183
OSDDDHPYSNZBPF-UHFFFAOYSA-N
1-pyrrolidinebutanenitrile,4-pyrrolidin-1-yl butanenitrile,1-pyrrolidinebutyronitrile,1-3-cyanopropyl pyrrolidine,4-1-pyrrolidine butyronitrile,1-pyrrolidinobutyronitrile,4-pyrrolidin-1-yl-butyronitrile,4-pyrrolidinylbutanenitrile,4-pyrrolidin-1-ylbutyronitrile,4-pyrrolidinobutyronitrile
118838
4-pyrrolidin-1-ylbutanenitrile
C1CCN(C1)CCCC#N
Flash Point 99°C
Specific Gravity 0.92
Boiling Point 115°C (18.0mmHg)
Refractive Index 1.4600 to 1.462
Color Brown to Colorless
Density 0.9200g/mL
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 138.21
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 1-Pyrrolidinebutanenitrile, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 252-612-4

RTECSNumber : UY0270000

TSCA : TSCA