Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pyrrolidinecarbodithioic acid ammonium salt, 98%, Thermo Scientific Chemicals

Catalog Number p-7027177
Click to view available options
Quantity:
25 g
100 g
5108-96-3
C5H8NS2-
146.246
MFCD00012720
VSWDORGPIHIGNW-UHFFFAOYSA-M
ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium
4311638
pyrrolidine-1-carbodithioate
C1CCN(C1)C(=S)[S-]
5108-96-3
C5H8NS2-
146.246
MFCD00012720
VSWDORGPIHIGNW-UHFFFAOYSA-M
ammonium pyrrolidyldithiocarbamate, apdc, ammonium 1-pyrrolidinecarbodithioate, ammonium tetramethylenedithiocarbamate, ammonium pyrrolidine-1-carbodithioate, ammonium 1-pyrrolidinyldithiocarbamate, ammonium 1-pyrrolidinedithiocarboxylate, 1-pyrrolidinecarbodithioic acid, ammonium salt, 1-pyrrolidinecarbodithioic acid ammonium salt, pyrrolidinedithiocarbamate ammonium
4311638
pyrrolidine-1-carbodithioate
C1CCN(C1)C(=S)[S-]

1-Pyrrolidinecarbodithioic acid ammonium salt is useful for biochemical applications including metal chelation. It is used to prevent the induction of nitric oxide synthase. It is also essential to inhibit apoptosis in leukemia HL-60 cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrrolidinecarbodithioic acid ammonium salt is useful for biochemical applications including metal chelation. It is used to prevent the induction of nitric oxide synthase. It is also essential to inhibit apoptosis in leukemia HL-60 cells.

Solubility
Soluble in water.

Notes
Hygroscopic. Air and light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 150°C to 155°C
Quantity 25 g
Beilstein 3730472
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 164.29
Percent Purity 98%
Chemical Name or Material 1-Pyrrolidinecarbodithioic acid ammonium salt
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-834-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit