Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pyrrolidino-1-cyclohexene, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
1125-99-1
C10H17N
151.25
MFCD00003163
KTZNVZJECQAMBV-UHFFFAOYSA-N
1-pyrrolidino-1-cyclohexene,1-pyrrolidinocyclohexene,1-cyclohex-1-en-1-yl pyrrolidine,pyrrolidine, 1-1-cyclohexen-1-yl,1-1-cyclohexen-1-yl pyrrolidine,1-1-pyrrolidinyl cyclohexene,cyclohexanone pyrrolidine enamine,1-pyrrolidinyl-1-cyclohexene,n-1-cyclohexenyl pyrrolidine,1-1-cyclohexenyl pyrrolidine
70768
1-(cyclohexen-1-yl)pyrrolidine
C1CCC(=CC1)N2CCCC2
1125-99-1
C10H17N
151.25
MFCD00003163
KTZNVZJECQAMBV-UHFFFAOYSA-N
1-pyrrolidino-1-cyclohexene,1-pyrrolidinocyclohexene,1-cyclohex-1-en-1-yl pyrrolidine,pyrrolidine, 1-1-cyclohexen-1-yl,1-1-cyclohexen-1-yl pyrrolidine,1-1-pyrrolidinyl cyclohexene,cyclohexanone pyrrolidine enamine,1-pyrrolidinyl-1-cyclohexene,n-1-cyclohexenyl pyrrolidine,1-1-cyclohexenyl pyrrolidine
70768
1-(cyclohexen-1-yl)pyrrolidine
C1CCC(=CC1)N2CCCC2
Boiling Point 114°C to 115°C (15.0mmHg)
Flash Point 66°C
Specific Gravity 0.94
Refractive Index 1.5210 to 1.523
Packaging Glass bottle
Infrared Spectrum Authentic
Density 0.9400g/mL
Quantity 25g
Color Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 151.25
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 1-Pyrrolidino-1-cyclohexene, 95%
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

EINECSNumber : 214-414-6

TSCA : TSCA