Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1'S,3S)-1-(1'-Phenylethyl)-5-oxo-3-pyrrolidine carboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
173340-19-7
C13H15NO3
233.267
MFCD03427553
CFGKWSDAMXTRHE-ONGXEEELSA-N
s-5-oxo-1-s-1-phenylethyl pyrrolidine-3-carboxylic acid,3s-5-oxo-1-1s-1-phenylethyl pyrrolidine-3-carboxylic acid,1's,3s---1-1'-phenylethyl-5-oxopyrrolidine-3-carboxylic acid,s-5-oxo-1-s-1-phenylethyl pyrrolidine-3-carboxylicacid,1's,3s-1-1'-phenylethyl-5-oxo-3-pyrrolidine carboxylic acid,3s-1-s-1-phenylethyl-5-oxo-3-pyrrolidinecarboxylic acid,1s,3s---1-1-phenylethyl-5-oxopyrrolidine-3-carboxylic acid,3-pyrrolidinecarboxylicacid, 5-oxo-1-1s-1-phenylethyl-, 3s,1 inverted exclamation markas,3s---1-1 inverted exclamation marka-phenylethyl-5-oxopyrrolidine-3-carboxylic acid
723630
(3S)-5-oxo-1-[(1S)-1-phenylethyl]pyrrolidine-3-carboxylic acid
CC(C1=CC=CC=C1)N2CC(CC2=O)C(=O)O
173340-19-7
C13H15NO3
233.267
MFCD03427553
CFGKWSDAMXTRHE-ONGXEEELSA-N
s-5-oxo-1-s-1-phenylethyl pyrrolidine-3-carboxylic acid,3s-5-oxo-1-1s-1-phenylethyl pyrrolidine-3-carboxylic acid,1's,3s---1-1'-phenylethyl-5-oxopyrrolidine-3-carboxylic acid,s-5-oxo-1-s-1-phenylethyl pyrrolidine-3-carboxylicacid,1's,3s-1-1'-phenylethyl-5-oxo-3-pyrrolidine carboxylic acid,3s-1-s-1-phenylethyl-5-oxo-3-pyrrolidinecarboxylic acid,1s,3s---1-1-phenylethyl-5-oxopyrrolidine-3-carboxylic acid,3-pyrrolidinecarboxylicacid, 5-oxo-1-1s-1-phenylethyl-, 3s,1 inverted exclamation markas,3s---1-1 inverted exclamation marka-phenylethyl-5-oxopyrrolidine-3-carboxylic acid
723630
(3S)-5-oxo-1-[(1S)-1-phenylethyl]pyrrolidine-3-carboxylic acid
CC(C1=CC=CC=C1)N2CC(CC2=O)C(=O)O
Melting Point 207°C to 211°C
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Rotation Condition − 104.00 (20.00°C c=2,MeOH)
Specific Rotation -100° to -108° (20°C, 589nm) (c=2, MeOH)
Quantity 1g
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 233.27
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material (1'S, 3S)-1-(1'-Phenylethyl)-5-oxo-3-pyrrolidine carboxylic acid, 98%
Show More Show Less