Learn More
Thermo Scientific™ 1-Step Heavy Protein IVT Kit
Express stable isotope-labeled (i.e., heavy) proteins by in vitro translation (IVT) for mass spectrometry using these cell-free expression kits.
$302.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PI88330
View Documents
Thermo Scientific™
88330
8 rxn kit Each for $302.00
Add to cart
PI88331
View Documents
Thermo Scientific™
88331
40 rxn kit This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
The Thermo Scientific™ 1-Step Heavy Protein In Vitro Protein Expression Kits enable rapid cell-free expression of recombinant proteins containing stable isotope-labeled (i.e., heavy) amino acids.
The 1-Step Heavy Protein IVT Kit uses a unique HeLa cell lysate supplemented with heavy amino acids for in vitro translation (IVT) of proteins with 90 to 95% isotope incorporation efficiency in less than 8 hours. Heavy proteins expressed using this system can be used as mass spectrometry controls for sample prep loss, digestion efficiency determination or as quantification standards.
Highlights:
  • Efficient – express heavy proteins with 90-95% stable isotope incorporation
  • Functional – uses the human translational machinery to express more biologically active proteins than other IVT systems
  • Flexible – express light proteins or heavy proteins containing 13C615N2 L-lysine and/or 13C615N4 L-arginine
  • Convenient – perform transcription and translation in a single step
  • High performance – greater yields compared to rabbit reticulocyte in vitro translation
Recommended for:
Identify ideal peptides for targeted quantitation; Verify protein digestion efficiency; Control for protein sample prep variability and affinity enrichment loss