Learn More
eMolecules​ 1-TERT-BUTYL-3-AZETIDINOL | 13156-04-2 | 1G | Purity: 95% DFS Item
Manufacturer:  eMolecules​ 248472853
AstaTech | 1-TERT-BUTYL-3-AZETIDINOL | 1G | 13156-04-2 | MFCD02323245
Catalog No. 50-157-8317
Due to product restrictions, please to purchase or view availability for this product.