Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butyl-4-piperidone, 97%, Thermo Scientific™

Catalog Number p-7032949
Click to view available options
Quantity:
250 mg
1 g
1465-76-5
C9H17NO
155.241
MFCD03411608
NKPMJJGLSFUOPK-UHFFFAOYSA-N
73840
1-tert-butylpiperidin-4-one
CC(C)(C)N1CCC(=O)CC1
1465-76-5
C9H17NO
155.241
MFCD03411608
NKPMJJGLSFUOPK-UHFFFAOYSA-N
73840
1-tert-butylpiperidin-4-one
CC(C)(C)N1CCC(=O)CC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 1-tert-butyl piperidin-4-one, 1-tert-butyl-piperidin-4-one, 1-tert-butyl-4-piperidone, 1-tert-butyl-4-piperidinone, 4-piperidinone,1-1,1-dimethylethyl, 4-piperidinone, 1-1,1-dimethylethyl, acmc-1bwol, n-t-butyl-4-piperidone, 1-t-butyl-4-piperidone, 1-tert-butyl-piperid-4-one
Formula Weight 155.24
Percent Purity 97%
Chemical Name or Material 1-tert-Butyl-4-piperidone
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit