Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butyldimethylsilyloxy)-2-propanone, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
Packaging:
Glass bottle
74685-00-0
C9H20O2Si
188.342
LHYDYTSJUGPFLA-UHFFFAOYSA-N
1-tert-butyldimethylsilyl oxy propan-2-one,1-tert-butyldimethylsilyloxy-2-propanone,acmc-1bhnq,tert-butyldimethylsiloxyacetone,t-butyl dimethyl silyloxy acetone,1-t-butyldimethylsilyloxy-2-propanone,1-tert-butyldimethylsilyloxy propan-2-one,1-tert-butyl dimethyl silyl oxy acetone,1-tert-butyl dimethyl-silyl oxy acetone
4124969
1-[tert-butyl(dimethyl)silyl]oxypropan-2-one
CC(=O)CO[Si](C)(C)C(C)(C)C
74685-00-0
C9H20O2Si
188.342
LHYDYTSJUGPFLA-UHFFFAOYSA-N
1-tert-butyldimethylsilyl oxy propan-2-one,1-tert-butyldimethylsilyloxy-2-propanone,acmc-1bhnq,tert-butyldimethylsiloxyacetone,t-butyl dimethyl silyloxy acetone,1-t-butyldimethylsilyloxy-2-propanone,1-tert-butyldimethylsilyloxy propan-2-one,1-tert-butyl dimethyl silyl oxy acetone,1-tert-butyl dimethyl-silyl oxy acetone
4124969
1-[tert-butyl(dimethyl)silyl]oxypropan-2-one
CC(=O)CO[Si](C)(C)C(C)(C)C
Quantity 5mL
Packaging Glass bottle
Color Colorless
Density 0.976g/mL
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 188.34
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1-(tert-Butyldimethylsilyloxy)-2-propanone
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.