Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-tert-Butylpiperazine, 97%

Click to view available options
Quantity:
1g
5g
38216-72-7
C8H18N2
142.246
MFCD00049359
PVMNSAIKFPWDQG-UHFFFAOYSA-N
n-tert-butylpiperazine,1-tert-butyl piperazine,n-t-butylpiperazine,1-t-butyl piperazine,1-tertbutyl piperazine,1-tert-butyl-piperazine,piperazine, 1-1,1-dimethylethyl,n-tert-butyl piperazine,tert-butylpiperazine,tert-butyl piperazine
3530572
1-tert-butylpiperazine
CC(C)(C)N1CCNCC1
38216-72-7
C8H18N2
142.246
MFCD00049359
PVMNSAIKFPWDQG-UHFFFAOYSA-N
n-tert-butylpiperazine,1-tert-butyl piperazine,n-t-butylpiperazine,1-t-butyl piperazine,1-tertbutyl piperazine,1-tert-butyl-piperazine,piperazine, 1-1,1-dimethylethyl,n-tert-butyl piperazine,tert-butylpiperazine,tert-butyl piperazine
3530572
1-tert-butylpiperazine
CC(C)(C)N1CCNCC1
Color Colorless to Yellow
Boiling Point 85°C to 87°C (22mmHg)
Melting Point 32°C to 35°C
Sensitivity Light, Air & Moisture Sensitive
Quantity 1g
UN Number UN3259
Formula Weight 142.24
Physical Form Low Melting Solid
Percent Purity 97%
Chemical Name or Material 1-tert-Butylpiperazine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.