Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Tetradecanol, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
112-72-1
C14H30O
214.39
MFCD00004757
HLZKNKRTKFSKGZ-UHFFFAOYSA-N
1-tetradecanol,myristyl alcohol,tetradecanol,tetradecyl alcohol,n-tetradecanol,myristic alcohol,n-tetradecyl alcohol,lanette k,loxanol v,lanette wax ks
8209
CHEBI:77417
tetradecan-1-ol
CCCCCCCCCCCCCCO
112-72-1
C14H30O
214.39
MFCD00004757
HLZKNKRTKFSKGZ-UHFFFAOYSA-N
1-tetradecanol,myristyl alcohol,tetradecanol,tetradecyl alcohol,n-tetradecanol,myristic alcohol,n-tetradecyl alcohol,lanette k,loxanol v,lanette wax ks
8209
CHEBI:77417
tetradecan-1-ol
CCCCCCCCCCCCCCO
Viscosity 6.4 mPa.s (60°C)
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 37.0°C to 40.0°C
Flash Point 145°C
Color White
Boiling Point 289.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)13OH
Beilstein 01, 428
Merck Index 15, 6421
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether, slightly soluble in alcohol
Formula Weight 214.39
Physical Form Low Melting Solid, Crystalline Powder, Crystals, Flakes, Pellets and/or Chunks
Percent Purity 99%
Chemical Name or Material 1-Tetradecanol
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 204-000-3