Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Tetradecylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
Packaging:
Glass bottle
2016-42-4
C14H31N
213.41
PLZVEHJLHYMBBY-UHFFFAOYSA-N
tetradecylamine,1-tetradecylamine,myristylamine,1-tetradecanamine,n-tetradecylamine,1-aminotetradecane,tetradecanamine,armeen 14,monotetradecylamine,alamine 5d
16217
tetradecan-1-amine
CCCCCCCCCCCCCCN
2016-42-4
C14H31N
213.41
PLZVEHJLHYMBBY-UHFFFAOYSA-N
tetradecylamine,1-tetradecylamine,myristylamine,1-tetradecanamine,n-tetradecylamine,1-aminotetradecane,tetradecanamine,armeen 14,monotetradecylamine,alamine 5d
16217
tetradecan-1-amine
CCCCCCCCCCCCCCN
Boiling Point 291°C
Refractive Index 1.5045 to 1.5065
Flash Point >110°C
Specific Gravity 0.993
Density 0.9930g/mL
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Melting Point 37°C to 42°C
Color Pink to White
Assay Percent Range 98%
Linear Formula CH3(CH2)13NH2
Beilstein 04, 201
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 213.41
Physical Form Crystals or Crystalline Solid
Percent Purity ≥97.5% (total base)
Chemical Name or Material 1-Tetradecylamine
Show More Show Less