Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Tetrahydro-2-furoyl)piperazine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
63074-07-7
C9H16N2O2
184.24
MFCD00190247
UKESBLFBQANJHH-UHFFFAOYSA-N
1-tetrahydro-2-furoyl piperazine,1-2-tetrahydrofuroyl piperazine,piperazin-1-yl tetrahydrofuran-2-yl methanone,1-2-tetrahydrofuroyl-piperazine,n-2-tetrahydrofuroyl piperazine,1-oxolane-2-carbonyl piperazine,1-tetrahydrofuran-2-ylcarbonyl piperazine,n-tetrahydro-2-furoyl piperazine,1-oxolan-2-ylcarbonyl piperazine,oxolan-2-yl piperazin-1-yl methanone
2734641
oxolan-2-yl(piperazin-1-yl)methanone
C1CC(OC1)C(=O)N2CCNCC2
63074-07-7
C9H16N2O2
184.24
MFCD00190247
UKESBLFBQANJHH-UHFFFAOYSA-N
1-tetrahydro-2-furoyl piperazine,1-2-tetrahydrofuroyl piperazine,piperazin-1-yl tetrahydrofuran-2-yl methanone,1-2-tetrahydrofuroyl-piperazine,n-2-tetrahydrofuroyl piperazine,1-oxolane-2-carbonyl piperazine,1-tetrahydrofuran-2-ylcarbonyl piperazine,n-tetrahydro-2-furoyl piperazine,1-oxolan-2-ylcarbonyl piperazine,oxolan-2-yl piperazin-1-yl methanone
2734641
oxolan-2-yl(piperazin-1-yl)methanone
C1CC(OC1)C(=O)N2CCNCC2
Refractive Index 1.5195 to 1.5215
Density 1.1700g/mL
Boiling Point 140°C (2.0 mbar)
Specific Gravity 1.17
Infrared Spectrum Authentic
Quantity 5g
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: miscible
Formula Weight 184.24
Physical Form Viscous Liquid
Percent Purity 98%
Chemical Name or Material 1-(Tetrahydro-2-furoyl)piperazine, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger