Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Thionaphthol, 99%

Click to view available options
Quantity:
1g
5g
25g
529-36-2
C10H8S
160.234
MFCD00039599
SEXOVMIIVBKGGM-UHFFFAOYSA-N
1-naphthalenethiol,1-thionaphthol,naphthalenethiol,1-naphthylthiol,unii-9a78cp495h,1-mercaptonaphthalene,.alpha.-naphthyl mercaptan,naphthalene thiol,1-napthalenethiol,naphthalen-1-thiol
68259
naphthalene-1-thiol
C1=CC=C2C(=C1)C=CC=C2S
529-36-2
C10H8S
160.234
MFCD00039599
SEXOVMIIVBKGGM-UHFFFAOYSA-N
1-naphthalenethiol,1-thionaphthol,naphthalenethiol,1-naphthylthiol,unii-9a78cp495h,1-mercaptonaphthalene,.alpha.-naphthyl mercaptan,naphthalene thiol,1-napthalenethiol,naphthalen-1-thiol
68259
naphthalene-1-thiol
C1=CC=C2C(=C1)C=CC=C2S
Applications
1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis.

Notes
Store away from air. Store under dry inert gas. Incompatible with air and oxidizing agents.
Quantity 5g
Formula Weight 160.24
Percent Purity 99%
Chemical Name or Material 1-Thionaphthol

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA