Alfa Aesar™ 1-(trans-2-Phenylethenyl)-4-(phenylethynyl)benzene, 97%

Manufacturer: Alfa Aesar™ H3063803

Quantity 1g

 View more versions of this product

Catalog No. AAH3063803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21850-30-6
    Molecular Formula C22H16
    Formula Weight 280.37
    MDL Number MFCD12407077
    Synonym 4-(trans-beta-Styryl)diphenylacetylene; 4-Ethynyl-trans-stilbene
    Chemical Name or Material 1-(trans-2-Phenylethenyl)-4-(phenylethynyl)benzene

    Specifications

    Formula Weight 280.37
    Molecular Formula C22H16
    MDL Number MFCD12407077
    CAS 21850-30-6
    Synonym 4-(trans-beta-Styryl)diphenylacetylene; 4-Ethynyl-trans-stilbene
    Melting Point 212° to 214°C
    Quantity 1g
    Chemical Name or Material 1-(trans-2-Phenylethenyl)-4-(phenylethynyl)benzene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit