Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Tridecene, 97%

Manufacturer:  Alfa Aesar™ B2441322

Quantity 100g

 View more versions of this product

Catalog No. AAB2441322


  Check Availability
Add to cart

Specifications

Description

Applications
It is used in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

2437-56-1
CH3(CH2)10CH=CH2
MFCD00008976
1-Tridecene
C13H26
182.35
1744660

Specifications

100g
C13H26
182.35
1744660
232° to 233°C
79°C (174°F)
97%
2437-56-1
CH3(CH2)10CH=CH2
MFCD00008976
Mild
−23°C
1-Tridecene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.

GHS P Statement
P301+P330+P331-P315
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.

Danger

RTECSNumber : YD4595000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit