Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Tridecene, 97%

Click to view available options
Quantity:
5g
25g
100g
2437-56-1
C13H26
182.351
MFCD00008976
VQOXUMQBYILCKR-UHFFFAOYSA-N
1-tridecene,n-tridec-1-ene,tridecylene,alpha-tridecene,undecylethylene,tridecene-1,unii-5b0u2s314c,ccris 5719,1-tridecylene,.alpha.-tridecene
17095
tridec-1-ene
CCCCCCCCCCCC=C
2437-56-1
C13H26
182.351
MFCD00008976
VQOXUMQBYILCKR-UHFFFAOYSA-N
1-tridecene,n-tridec-1-ene,tridecylene,alpha-tridecene,undecylethylene,tridecene-1,unii-5b0u2s314c,ccris 5719,1-tridecylene,.alpha.-tridecene
17095
tridec-1-ene
CCCCCCCCCCCC=C
Applications
It is used in pharmaceutical industry.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
Boiling Point 232°C to 233°C
Odor Mild
Flash Point 79°C (174°F)
Quantity 5g
Melting Point -23°C
Linear Formula CH3(CH2)10CH=CH2
Beilstein 1744660
Formula Weight 182.35
Percent Purity 97%
Chemical Name or Material 1-Tridecene

GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.

GHS P Statement
P301+P330+P331-P315
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Get immediate medical advice/attention.

Danger

RTECSNumber : YD4595000

TSCA : TSCA