Alfa Aesar™ 1-(Trifluoromethyl)vinyl acetate, 97%

Manufacturer: Alfa Aesar™ B2213330

Quantity 250g

 View more versions of this product

Catalog No. AAB2213330

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2247-91-8
    Molecular Formula C5H5F3O2
    Formula Weight 154.09
    MDL Number MFCD00040844
    UN Number UN3272
    Beilstein 2078331
    Synonym 1-(Trifluoromethyl)ethenyl acetate; 3,3,3-Trifluoropropen-2-yl acetate
    Chemical Name or Material 1-(Trifluoromethyl)vinyl acetate

    Specifications

    Formula Weight 154.09
    TSCA TSCA
    Odor Pungent
    UN Number UN3272
    Synonym 1-(Trifluoromethyl)ethenyl acetate; 3,3,3-Trifluoropropen-2-yl acetate
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
    Flash Point 2°C (35°F)
    Molecular Formula C5H5F3O2
    MDL Number MFCD00040844
    CAS 2247-91-8
    Beilstein 2078331
    Health Hazard 1 Danger
    Boiling Point 86° to 88°C
    Quantity 250g
    Chemical Name or Material 1-(Trifluoromethyl)vinyl acetate
    Assay 97%

    Description

    Applications
    1-(Trifluoromethyl)vinyl acetate is used as a chemical, organic Intermediates.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Moisture sensitive. Store away from strong oxidizing agents, heat, water/ moisture.

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit