Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-(Trimethylsiloxy)cyclohexene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
Specifications
Boiling Point | 165°C |
Quantity | 10 g |
UN Number | UN1993 |
Beilstein | 1859394 |
Flash Point | 42°C (107°F) |
Solubility Information | Not miscible or difficult to mix. |
Formula Weight | 170.33 |
Percent Purity | 98% |
Refractive Index | 1.447 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H226 |
Hazard Statement | GHS H Statement H226 Flammable liquid and vapour. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
EINECSNumber | 229-675-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction