Alfa Aesar™ 1-Trimethylsilyl-1,4-pentadiyne, 98%

Manufacturer: Alfa Aesar™ H5343606

Quantity 5g

 View more versions of this product

Catalog No. AAH5343606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 71789-10-1
    Molecular Formula C8H12Si
    Linear Formula HC≡CCH2C≡CSi(CH3)3
    Formula Weight 136.27
    MDL Number MFCD00066350
    UN Number UN1993
    Chemical Name or Material 1-Trimethylsilyl-1,4-pentadiyne

    Specifications

    Formula Weight 136.27
    Molecular Formula C8H12Si
    UN Number UN1993
    Health Hazard 1 Danger
    Linear Formula HC≡CCH2C≡CSi(CH3)3
    MDL Number MFCD00066350
    CAS 71789-10-1
    Boiling Point 42°C (25mmHg)
    Quantity 5g
    Flash Point 8°C (46°F)
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 1-Trimethylsilyl-1,4-pentadiyne
    Assay 98%

    Description

    Notes
    Incompatible with strong oxidizing agents and ammonia.

    GHS H Statement
    H225
    Highly flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit