Alfa Aesar™ 1-Trimethylsilyl-1-propyne, 98%

Manufacturer: Alfa Aesar™ B2487503

Quantity 1g

 View more versions of this product

Catalog No. AAB2487503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6224-91-5
    Molecular Formula CH3C≡CSi(CH3)3
    Formula Weight 112.25
    MDL Number MFCD00009271
    UN Number UN1993
    Beilstein 1071311
    Synonym Methyl trimethylsilyl acetylene; Propargyltrimethylsilane
    Chemical Name or Material 1-Trimethylsilyl-1-propyne

    Specifications

    Formula Weight 112.25
    Molecular Formula CH3C≡CSi(CH3)3
    MDL Number MFCD00009271
    CAS 6224-91-5
    UN Number UN1993
    Beilstein 1071311
    Synonym Methyl trimethylsilyl acetylene; Propargyltrimethylsilane
    Boiling Point 99° to 100°C
    Melting Point −69°C
    Quantity 1g
    Flash Point −3°C (26°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapour.
    Health Hazard 3 GHS P Statement
    P210-P243-P403
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Take precautionary measures against static discharge.
    Store in a well-ventilated place.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 1-Trimethylsilyl-1-propyne
    Assay 98%

    Description

    Applications
    1-Trimethylsilyl-1-propyne is employed in the preparation of alkynyl silane derivatives. It is also used to prepare highly substituted indenes using palladium-catalyzed carboannulation. Further, it plays an important role in the preparation of indenones through a rhodium-catalyzed reaction with 2-bromophenylboronic acids.

    Solubility
    Miscible with toluene.

    Notes
    Store in a cool place. Incompatible with strong oxidizing agents and strong bases.

    GHS H Statement
    H225
    Highly flammable liquid and vapour.

    GHS P Statement
    P210-P243-P403
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Take precautionary measures against static discharge.
    Store in a well-ventilated place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit