Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1-Tritylimidazole-4-carboxaldehyde, 98%
Click to view available options
Quantity:
1g
5g
25g
Description
Applications
1-Tritylimidazole-4-carboxaldehyde is used in the synthesis of histamine and girolline
Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agents, air.
1-Tritylimidazole-4-carboxaldehyde is used in the synthesis of histamine and girolline
Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agents, air.
Specifications
Sensitivity | Air Sensitive |
Quantity | 5g |
Melting Point | ~192°C (decomposition) |
Formula Weight | 338.41 |
Percent Purity | 98% |
Chemical Name or Material | 1-Tritylimidazole-4-carboxaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures