Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Undecanol 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
112-42-5
C11H24O
172.31
MFCD00004751
KJIOQYGWTQBHNH-UHFFFAOYSA-N
1-undecanol,undecyl alcohol,undecanol,n-undecanol,n-undecyl alcohol,hendecanoic alcohol,hendecyl alcohol,1-hendecanol,n-hendecylenic alcohol,tip-nip
8184
CHEBI:87499
undecan-1-ol
CCCCCCCCCCCO
112-42-5
C11H24O
172.31
MFCD00004751
KJIOQYGWTQBHNH-UHFFFAOYSA-N
1-undecanol,undecyl alcohol,undecanol,n-undecanol,n-undecyl alcohol,hendecanoic alcohol,hendecyl alcohol,1-hendecanol,n-hendecylenic alcohol,tip-nip
8184
CHEBI:87499
undecan-1-ol
CCCCCCCCCCCO
Quantity 100mL
Refractive Index 1.4370 to 1.4420
Flash Point >112°C
Melting Point 11.0°C
Density 0.8310g/mL
Boiling Point 146.0°C (30.0 mmHg)
Packaging Glass bottle
Specific Gravity 0.831
Color Colorless to Yellow
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)10OH
Beilstein 01, 427
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in 60% alcohol
Formula Weight 172.31
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Undecanol, 98%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 203-970-5

RTECSNumber : YQ3155000

TSCA : TSCA