Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10.00 pH Buffer Products for Science Education

Buffer, 10.00 pH; For meters w/0.01 resolution; Standardized against NIST-certified pH references within 0.01 pH at 77°F (25°C) 500mL bottle

Catalog No. S282891

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • pH buffer solutions allow the user to adjust their system to read accurate measurements
  • Store buffers away from direct sunlight.
Specifications

Specifications

Buffer
10.00 pH Buffer
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit