Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 10,12-Docosadiynedioic acid, 95%
Click to view available options
Quantity:
1g
5g
Description
Applications
Polymerizable diacid (10,12-docosadiynedioic acid) was used as obtained from GFS Chemicals. Synthesis of dimesogenic liquid crystals with both cholesterol and azobenzene groups is by the esterification of 10,12-docosadiynedioic acid. .
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Polymerizable diacid (10,12-docosadiynedioic acid) was used as obtained from GFS Chemicals. Synthesis of dimesogenic liquid crystals with both cholesterol and azobenzene groups is by the esterification of 10,12-docosadiynedioic acid. .
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
Sensitivity | Light sensitive |
Melting Point | 110°C to 112°C |
Quantity | 1g |
Beilstein | 1803395 |
Solubility Information | Insoluble in water. |
Formula Weight | 362.51 |
Percent Purity | 95% |
Chemical Name or Material | 10,12-Docosadiynedioic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures