Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10,12-Tricosadiynoic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
100mg
1g
66990-30-5
C23H38O2
346.555
MFCD00041683
DIEDVCMBPCRJFQ-UHFFFAOYSA-N
10,12-tricosadiynoic acid,10,12-tricosadiynoicacid,acmc-209nx8,10,12-tricosadiynoic acid gc
538457
tricosa-10,12-diynoic acid
CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
66990-30-5
C23H38O2
346.555
MFCD00041683
DIEDVCMBPCRJFQ-UHFFFAOYSA-N
10,12-tricosadiynoic acid,10,12-tricosadiynoicacid,acmc-209nx8,10,12-tricosadiynoic acid gc
538457
tricosa-10,12-diynoic acid
CCCCCCCCCCC#CC#CCCCCCCCCC(=O)O
Color White-Yellow
Melting Point 59°C
Quantity 100mg
Formula Weight 346.56
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 10,12-Tricosadiynoic Acid

TSCA : No

Recommended Storage : Refrigerator