Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 10-Chloro-3-decyne, 98%
Click to view available options
Quantity:
5g
25g
Specifications
Quantity | 5g |
Refractive Index | 1.462 |
Density | 0.92 |
Boiling Point | 122°C (20mmHg) |
Flash Point | >110°C (230°F) |
Formula Weight | 172.7 |
Percent Purity | 98% |
Chemical Name or Material | 10-Chloro-3-decyne |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures