Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 10-Chloro-3-decyne, 98%

Click to view available options
Quantity:
5g
25g
18295-64-2
C10H17Cl
172.696
MFCD01321154
BQQIODGRFRSXTM-UHFFFAOYSA-N
10-chloro-3-decyne,3-decyne, 10-chloro,chlorodecyne,7-decyn-1-yl chloride,acmc-209ek5,bqqiodgrfrsxtm-uhfffaoysa
87561
10-chlorodec-3-yne
CCC#CCCCCCCCl
18295-64-2
C10H17Cl
172.696
MFCD01321154
BQQIODGRFRSXTM-UHFFFAOYSA-N
10-chloro-3-decyne,3-decyne, 10-chloro,chlorodecyne,7-decyn-1-yl chloride,acmc-209ek5,bqqiodgrfrsxtm-uhfffaoysa
87561
10-chlorodec-3-yne
CCC#CCCCCCCCl
Quantity 25g
Flash Point >110°C (230°F)
Boiling Point 122°C (20mmHg)
Formula Weight 172.7
Percent Purity 98%
Chemical Name or Material 10-Chloro-3-decyne