Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 10-Deacetylbaccatin III, 95%

Click to view available options
Quantity:
5mg
25mg
32981-86-5
C29H36O10
544.597
MFCD00132913
YWLXLRUDGLRYDR-ZUKOLHLZSA-N
129316875
CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1O)O)OC(=O)C5=CC=CC=C5)(CO4)OC(=O)C)O)C)O
32981-86-5
C29H36O10
544.597
MFCD00132913
YWLXLRUDGLRYDR-ZUKOLHLZSA-N
129316875
CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1O)O)OC(=O)C5=CC=CC=C5)(CO4)OC(=O)C)O)C)O

An intermediate in the synthesis of paclitaxel

Melting Point 229°C to 236°C
Boiling Point 717°C
Quantity 25mg
Color White
Solubility Information Soluble in chloroform; Also soluble in methanol with warming; Insoluble in water
Formula Weight 544.59
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 10-Deacetylbaccatin III

storageNote1 : 2° to 8°C