Alfa Aesar™ 10-Isocyanatodecyltrimethoxysilane, 95%

Manufacturer: Alfa Aesar™ H3355503

Quantity 1g

 View more versions of this product

Catalog No. AAH3355503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1310372-82-7
    Molecular Formula C14H29NO4Si
    Linear Formula OCN(CH2)10Si(OCH3)3
    Formula Weight 303.47
    MDL Number MFCD19105613
    UN Number UN2810
    Chemical Name or Material 10-Isocyanatodecyltrimethoxysilane

    Specifications

    Formula Weight 303.47
    MDL Number MFCD19105613
    Boiling Point 120° to 125°C (0.1mmHg)
    Health Hazard 2 GHS H Statement
    H331-H318-H302-H312-H315-H335
    Toxic if inhaled.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C14H29NO4Si
    Linear Formula OCN(CH2)10Si(OCH3)3
    CAS 1310372-82-7
    UN Number UN2810
    Quantity 1g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material 10-Isocyanatodecyltrimethoxysilane
    Assay 95%

    GHS H Statement
    H331-H318-H302-H312-H315-H335
    Toxic if inhaled.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit