Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10-Undecen-1-ol, 98+%, stabilized, Acros Organics

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
112-43-6
C11H22O
170.29
MFCD00004750
GIEMHYCMBGELGY-UHFFFAOYSA-N
10-undecen-1-ol,undecylenic alcohol,undecylenyl alcohol,10-undecenol,1-undecen-11-ol,10-undecylen-1-ol,11-hydroxy-1-undecene,omega-undecenyl alcohol,undecen-1-ol,.omega.-undecenyl alcohol
8185
undec-10-en-1-ol
C=CCCCCCCCCCO
112-43-6
C11H22O
170.29
MFCD00004750
GIEMHYCMBGELGY-UHFFFAOYSA-N
10-undecen-1-ol,undecylenic alcohol,undecylenyl alcohol,10-undecenol,1-undecen-11-ol,10-undecylen-1-ol,11-hydroxy-1-undecene,omega-undecenyl alcohol,undecen-1-ol,.omega.-undecenyl alcohol
8185
undec-10-en-1-ol
C=CCCCCCCCCCO
Melting Point -3.0°C
Boiling Point 245.0°C to 248.0°C
Infrared Spectrum Authentic
Specific Gravity 0.846
Density 0.8460g/mL
Quantity 100g
Packaging Glass bottle
Refractive Index 1.4460 to 1.4530
Color Colorless
Flash Point 93°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Linear Formula H2C=CH(CH2)9OH
Beilstein 01,452
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 170.29
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material 10-Undecen-1-ol
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 203-971-