Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gosselin™ 100 x 15mm Polystyrene Petri Dishes

Ideal for microbiology examination either by surface plating or plate pouring techniques

Manufacturer:  Gosselin™ BP94A01

 View more versions of this product

Catalog No. 07-202-010


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Yes
15 mm
500
Yes
100 mm
Polystyrene
100 x 15mm (3.9 x 0.05 in.)
Automation Petri Dishes
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit