Learn More
Sino Biological Human EGFR / HER1 / ErbB1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10001H08H20
A DNA sequence encoding the extracellular domain (Met 1-Ser 645) of human EGFR (NP_005219) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-3724