Learn More
Sino Biological EGFR / HER1 / ErbB1 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10001MM01100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EGFR / ErbB / HER1 (rh EGFR / ErbB / HER1; Catalog #10001-H08H; Met 1-Ser 645; NP_005219). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0246