Learn More
Sino Biological EGFR / HER1 / ErbB1 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10001MM0850
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EGFR / HER1 / ErbB1 (rh EGFR / HER1 / ErbB1; Catalog#10001-H08H; NP_005219; Met1-Ser645). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0251