Learn More
Sino Biological EGFR / HER1 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10001RE11100
This antibody was obtained from a rabbit immunized with purified, recombinant Human EGFR / HER1 (rh EGFR / HER1; Catalog#10001-H08H; NP_005219; Met1-Ser645).
Catalog No. 50-161-0277